accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175291-56-2

IUPAC Name: (2S,3R)-3-{[(benzyloxy)(hydroxy)phosphoryl]oxy}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: (2S,3R)-3-{[benzyloxy(hydroxy)phosphoryl]oxy}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid|FMOC-O-(BENZYLPHOSPHO)-L-THREONINE
Molecular Weight (g/mol): 511.47
Molecular Formula: C26H26NO8P
InChi Key: HOFDVXHILSPFNS-OSPHWJPCSA-N
SMILES: C[C@@H](OP(O)(=O)OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)