accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClNOOBr

CAS RN 175334-69-7

IUPAC Name: 2-bromo-1-[3-(2,4-dichlorophenyl)-1,2-oxazol-5-yl]ethan-1-one
Synonyms: 2-bromo-1-[3-(2,4-dichlorophenyl)-1,2-oxazol-5-yl]ethanone
Molecular Weight (g/mol): 334.98
Molecular Formula: C11H6BrCl2NO2
InChi Key: WYJZTLHKOJBVSA-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(C=C1)C1=NOC(=C1)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results