accessibility menu, dialog, popup

UserName

Viewing profile as user:
SHSH

CAS RN 17534-15-5

IUPAC Name: benzene-1,2-dithiol
Synonyms: 1,2-benzenedithiol
Molecular Weight (g/mol): 142.23
Molecular Formula: C6H6S2
InChi Key: JRNVQLOKVMWBFR-UHFFFAOYSA-N
SMILES: SC1=C(S)C=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)