accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175357-73-0

IUPAC Name: 4-bromo-N-[(4-methoxyphenyl)methyl]aniline
Synonyms: 4-bromo-N-[(4-methoxyphenyl)methyl]aniline
Molecular Weight (g/mol): 292.18
Molecular Formula: C14H14BrNO
InChi Key: LJKLYFDMLGYRCA-UHFFFAOYSA-N
SMILES: COC1=CC=C(CNC2=CC=C(Br)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results