accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NHNCH3H3C

CAS RN 17536-00-4

IUPAC Name: 3,5-bis(propan-2-yl)-1H-pyrazole
Synonyms: 3,5-diisopropyl-1H-pyrazole
Molecular Weight (g/mol): 152.24
Molecular Formula: C9H16N2
InChi Key: PJSJVTZSFIRYIZ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(=NN1)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results