CAS RN 175361-81-6
IUPAC Name:
4,4,5,5-tetramethyl-2-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]-1,3,2-dioxaborolane
Synonyms:
4,4,5,5-tetramethyl-2-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol):
336.06
Molecular Formula:
C16H26B2O4S
InChi Key:
AOJXAKMKFDBHHT-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC=C(S1)B1OC(C)(C)C(C)(C)O1