accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1754-49-0

IUPAC Name: diethyl phenylphosphonate
Synonyms: diethyl phenylphosphonate
Molecular Weight (g/mol): 214.2
Molecular Formula: C10H15O3P
InChi Key: VZEGPPPCKHRYGO-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results