accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONON

CAS RN 175463-32-8

IUPAC Name: tert-butyl 3-cyano-4-oxopyrrolidine-1-carboxylate
Synonyms: tert-butyl 3-cyano-4-oxopyrrolidine-1-carboxylate
Molecular Weight (g/mol): 210.23
Molecular Formula: C10H14N2O3
InChi Key: RKUQLKRPWWYRIG-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(C#N)C(=O)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)