accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOH3CCH3

CAS RN 1755-15-3

IUPAC Name: 2-acetyl-5,5-dimethylcyclohexane-1,3-dione
Synonyms: 2-acetyl-5,5-dimethylcyclohexane-1,3-dione
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: ITSKWKZDPHAQNK-UHFFFAOYSA-N
SMILES: CC(=O)C1C(=O)CC(C)(C)CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results