accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17560-51-9

IUPAC Name: 7-chloro-2-methyl-3-(2-methylphenyl)-4-oxo-1,2,3,4-tetrahydroquinazoline-6-sulfonamide
Synonyms: metolazone metolazona metolazone (product) metolazone preparation metolazone (substance) metalazone xuret zaroxolyn microx metalozone
Molecular Weight (g/mol): 365.83
Molecular Formula: C16H16ClN3O3S
InChi Key: AQCHWTWZEMGIFD-UHFFFAOYNA-N
SMILES: CC1NC2=CC(Cl)=C(C=C2C(=O)N1C1=CC=CC=C1C)S(N)(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results