accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175696-70-5

IUPAC Name: 1-[3-(pyrrolidin-1-yl)phenyl]methanamine
Synonyms: 1-[3-(pyrrolidin-1-yl)phenyl]methanamine
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: BXTWTBYGOKOWQX-UHFFFAOYSA-N
SMILES: NCC1=CC(=CC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results