accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOONH2

CAS RN 1758-68-5

IUPAC Name: 1,2-diamino-9,10-dihydroanthracene-9,10-dione
Synonyms: 1,2-diaminoanthracene-9,10-dione
Molecular Weight (g/mol): 238.25
Molecular Formula: C14H10N2O2
InChi Key: LRMDXTVKVHKWEK-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results