accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17584-12-2

IUPAC Name: dimethyl-1,2,4-triazin-3-amine
Synonyms: dimethyl-1,2,4-triazin-3-amine
Molecular Weight (g/mol): 124.15
Molecular Formula: C5H8N4
InChi Key: UIKGLXJNZXSPGV-UHFFFAOYSA-N
SMILES: CC1=NN=C(N)N=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results