accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1759-53-1

IUPAC Name: cyclopropanecarboxylic acid
Synonyms: cyclopropanecarboxylic acid
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: YMGUBTXCNDTFJI-UHFFFAOYSA-N
SMILES: OC(=O)C1CC1
Boiling Point 
  • (3)
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (6)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)