accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17594-47-7

IUPAC Name: barium(2+) bis(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Synonyms: barium(2+) bis(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Molecular Weight (g/mol): 503.87
Molecular Formula: C22H38BaO4
InChi Key: VJRLFEOHKCDHBT-UHFFFAOYSA-N
SMILES: [Ba++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results