accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175968-39-5

IUPAC Name: 2,3-difluoro-4-hydroxybenzoic acid
Synonyms: 2,3-difluoro-4-hydroxybenzoic acid
Molecular Weight (g/mol): 174.1
Molecular Formula: C7H4F2O3
InChi Key: XIZIDHMVDRRFBT-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(F)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results