accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17606-31-4

IUPAC Name: {1,3-bis[(benzenesulfonyl)sulfanyl]propan-2-yl}dimethylamine
Synonyms: ruban ZZ-doricida bancol victenon nereistoxin dibenzenesulfonate bensultap ZZ-doricida insecticide
Molecular Weight (g/mol): 431.6
Molecular Formula: C17H21NO4S4
InChi Key: YFXPPSKYMBTNAV-UHFFFAOYSA-N
SMILES: CN(C)C(CSS(=O)(=O)C1=CC=CC=C1)CSS(=O)(=O)C1=CC=CC=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results