accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17608-10-5

ClNCS

CAS RN 17608-10-5

IUPAC Name: 1-chloro-4-(2-isothiocyanatoethyl)benzene
Synonyms: 1-chloro-4-(2-isothiocyanatoethyl)benzene
Molecular Weight (g/mol): 197.68
Molecular Formula: C9H8ClNS
InChi Key: MRJJYUJULSZFDV-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CCN=C=S)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results