accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3COOH2N

CAS RN 17609-48-2

IUPAC Name: hydrogen ethyl 2-(2-aminophenyl)acetate chloride
Synonyms: hydrogen ethyl 2-(2-aminophenyl)acetate chloride
Molecular Weight (g/mol): 215.68
Molecular Formula: C10H14ClNO2
InChi Key: QITKMRXHUSYVSD-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCOC(=O)CC1=CC=CC=C1N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)