accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17609-52-8

IUPAC Name: (2R)-2-{[(benzyloxy)carbonyl]amino}-2-phenylacetic acid
Synonyms: (R)-{[(benzyloxy)carbonyl]amino}(phenyl)acetic acid
Molecular Weight (g/mol): 285.3
Molecular Formula: C16H15NO4
InChi Key: RLDJWBVOZVJJOS-CQSZACIVSA-N
SMILES: OC(=O)[C@H](NC(=O)OCC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)