accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OHCl

CAS RN 17609-80-2

IUPAC Name: 4-amino-3-chlorophenol
Synonyms: 4-amino-3-chlorophenol
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: PNLPXABQLXSICH-UHFFFAOYSA-N
SMILES: NC1=CC=C(O)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results