accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHBrO

CAS RN 1761-61-1

IUPAC Name: 5-bromo-2-hydroxybenzaldehyde
Synonyms: 5-bromo-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 201.02
Molecular Formula: C7H5BrO2
InChi Key: MKKSTJKBKNCMRV-UHFFFAOYSA-N
SMILES: OC1=CC=C(Br)C=C1C=O
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (5)
  • (2)
Quantity 
  • (3)
  • (4)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (4)
  • (2)
  • (5)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)