accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1762-84-1

IUPAC Name: 4-bromo-4'-phenyl-1,1'-biphenyl
Synonyms: 4-bromo-4'-phenyl-1,1'-biphenyl
Molecular Weight (g/mol): 309.21
Molecular Formula: C18H13Br
InChi Key: XNBGHWRYLJOQAU-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results