accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOHSOOOO

CAS RN 17625-03-5

IUPAC Name: sodium 3-sulfobenzoate
Synonyms: sodium 3-sulfobenzoate
Molecular Weight (g/mol): 224.16
Molecular Formula: C7H5NaO5S
InChi Key: KQHKITXZJDOIOD-UHFFFAOYSA-M
SMILES: [Na+].OS(=O)(=O)C1=CC=CC(=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)