CAS RN 1763-23-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Verispec™ PFAS Standards, Certified Reference Material
Manufactured and tested in an ISO 17025 and 17034 accredited facility for applications in environmental testing, food safety, water quality, and more.