accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3Cl

CAS RN 17639-93-9

IUPAC Name: methyl 2-chloropropanoate
Synonyms: methyl 2-chloropropanoate
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: JLEJCNOTNLZCHQ-UHFFFAOYNA-N
SMILES: COC(=O)C(C)Cl
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)