accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO2H2H2H2H2H2H2H2H

CAS RN 17647-74-4

IUPAC Name: (²H₈)-1,4-dioxane
Synonyms: (²H₈)-1,4-dioxane
Molecular Weight (g/mol): 96.16
Molecular Formula: C4H8O2
InChi Key: RYHBNJHYFVUHQT-SVYQBANQSA-N
SMILES: [2H]C1([2H])OC([2H])([2H])C([2H])([2H])OC1([2H])[2H]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results