CAS RN 1765-40-8
2,3,4,5,6-Pentafluorobenzyl bromide, 97%
CAS: 1765-40-8 Molecular Formula: C7H2BrF5 Molecular Weight (g/mol): 260.989 MDL Number: MFCD00000299 InChI Key: XDEPVFFKOVDUNO-UHFFFAOYSA-N Synonym: 2,3,4,5,6-pentafluorobenzyl bromide,pentafluorobenzyl bromide,1-bromomethyl-2,3,4,5,6-pentafluorobenzene,1-bromomethylpentafluorobenzene,alpha-bromo-2,3,4,5,6-pentafluorotoluene,bromomethyl pentafluorobenzene,benzene, bromomethyl pentafluoro,2,3,4,5,6-pentafluorobenzylbromide,pfbbr,pentafluorobenzylbromide PubChem CID: 74484 IUPAC Name: 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene SMILES: C(C1=C(C(=C(C(=C1F)F)F)F)F)Br
alpha-Bromo-2,3,4,5,6-pentafluorotoluene, 97%
CAS: 1765-40-8 MDL Number: MFCD00000299 InChI Key: XDEPVFFKOVDUNO-UHFFFAOYSA-N Synonym: 2,3,4,5,6-pentafluorobenzyl bromide,pentafluorobenzyl bromide,1-bromomethyl-2,3,4,5,6-pentafluorobenzene,1-bromomethylpentafluorobenzene,alpha-bromo-2,3,4,5,6-pentafluorotoluene,bromomethyl pentafluorobenzene,benzene, bromomethyl pentafluoro,2,3,4,5,6-pentafluorobenzylbromide,pfbbr,pentafluorobenzylbromide PubChem CID: 74484 IUPAC Name: 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene SMILES: C(C1=C(C(=C(C(=C1F)F)F)F)F)Br
LiChropur™ 2,3,4,5,6-Pentafluorobenzyl bromide, ≥98.5% (GC), MilliporeSigma™ Supelco™
MDL Number: MFCD00000299 Synonym: PFB-Br; alpha-Bromo-2,3,4,5,6-pentafluorotoluene; (Bromomethyl)pentafluorobenzene
Pentafluorobenzyl Bromide 98.0+%, TCI America™
CAS: 1765-40-8 Molecular Formula: C7H2BrF5 Molecular Weight (g/mol): 260.989 MDL Number: MFCD00000299 InChI Key: XDEPVFFKOVDUNO-UHFFFAOYSA-N Synonym: 2,3,4,5,6-pentafluorobenzyl bromide,pentafluorobenzyl bromide,1-bromomethyl-2,3,4,5,6-pentafluorobenzene,1-bromomethylpentafluorobenzene,alpha-bromo-2,3,4,5,6-pentafluorotoluene,bromomethyl pentafluorobenzene,benzene, bromomethyl pentafluoro,2,3,4,5,6-pentafluorobenzylbromide,pfbbr,pentafluorobenzylbromide PubChem CID: 74484 IUPAC Name: 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene SMILES: C(C1=C(C(=C(C(=C1F)F)F)F)F)Br
Sigma Aldrich 2,3,4,5,6-Pentafluorobenzyl bromide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
a-Bromo-2,3,4,5,6-Pentafluorotoluene, SPEX CertiPrep™
CAS: 1765-40-8 Molecular Formula: C7H2BrF5 Molecular Weight (g/mol): 260.989 InChI Key: XDEPVFFKOVDUNO-UHFFFAOYSA-N PubChem CID: 74484 IUPAC Name: 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene SMILES: C(C1=C(C(=C(C(=C1F)F)F)F)F)Br