accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClOPClOOClClCl

CAS RN 17672-53-6

IUPAC Name: bis(2,2,2-trichloroethyl) phosphorochloridate
Synonyms: bis(2,2,2-trichloroethyl) phosphorochloridate
Molecular Weight (g/mol): 379.2
Molecular Formula: C4H4Cl7O3P
InChi Key: ZHHCWQGVXYGWCW-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)COP(Cl)(=O)OCC(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results