accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17673-68-6

IUPAC Name: 1,2-dimethyl (1R,2R)-cyclohex-4-ene-1,2-dicarboxylate
Synonyms: 1,2-dimethyl (1R,2R)-cyclohex-4-ene-1,2-dicarboxylate
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: DVVAGRMJGUQHLI-HTQZYQBOSA-N
SMILES: COC(=O)[C@@H]1CC=CC[C@H]1C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results