accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 176787-96-5

IUPAC Name: 5'-bromo-[2,2'-bithiophene]-5-carbonitrile
Synonyms: 5'-bromo-[2,2'-bithiophene]-5-carbonitrile
Molecular Weight (g/mol): 270.16
Molecular Formula: C9H4BrNS2
InChi Key: OHVCHDZJRUEOKH-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=CC=C(S1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results