accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OH3COH3COCH3OCH3

CAS RN 176798-33-7

IUPAC Name: 2,2,3,3-tetramethoxybutane
Synonyms: 2,2,3,3-tetramethoxybutane
Molecular Weight (g/mol): 178.23
Molecular Formula: C8H18O4
InChi Key: COEBCOIFCLAOFZ-UHFFFAOYSA-N
SMILES: COC(C)(OC)C(C)(OC)OC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)