accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17685-52-8

COCOCOCOCOCOCOCOCOCOCOCOFeFeFe

CAS RN 17685-52-8

IUPAC Name: dodecakis(methanidylidyneoxidanium) triiron
Synonyms: dodecakis(carbon monoxide) triiron
Molecular Weight (g/mol): 503.66
Molecular Formula: C12Fe3O12
InChi Key: HLYRMDDXFDINCB-UHFFFAOYSA-N
SMILES: [Fe].[Fe].[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)