accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17687-47-7

IUPAC Name: 1-[4-(methylamino)phenyl]ethan-1-one
Synonyms: 1-[4-(methylamino)phenyl]ethanone
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: GDBAYICEBDZUNM-UHFFFAOYSA-N
SMILES: CNC1=CC=C(C=C1)C(C)=O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results