accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17697-12-0

IUPAC Name: benzeneseleninyl benzeneseleninate
Synonyms: benzeneseleninyl benzeneseleninate
Molecular Weight (g/mol): 360.15
Molecular Formula: C12H10O3Se2
InChi Key: FHPZOWOEILXXBD-UHFFFAOYSA-N
SMILES: O=[Se](O[Se](=O)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results