accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3ON

CAS RN 176977-56-3

IUPAC Name: 4-[6-methoxy-2-(4-methoxyphenyl)-1-benzofuran-3-carbonyl]benzonitrile
Synonyms: 4-[6-methoxy-2-(4-methoxyphenyl)-1-benzofuran-3-carbonyl]benzonitrile
Molecular Weight (g/mol): 383.4
Molecular Formula: C24H17NO4
InChi Key: RYNSGDFWBJWWSZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=C(C(=O)C2=CC=C(C=C2)C#N)C2=CC=C(OC)C=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results