accessibility menu, dialog, popup

UserName

Viewing profile as user:
BSiCH3CH3H3CHOOH

CAS RN 177171-16-3

IUPAC Name: [3-(trimethylsilyl)phenyl]boronic acid
Synonyms: 3-(trimethylsilyl)phenylboronic acid
Molecular Weight (g/mol): 194.11
Molecular Formula: C9H15BO2Si
InChi Key: REMKRZLFPLDTKR-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results