accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOH3COOCH3

CAS RN 17720-63-7

IUPAC Name: N-acetylacetamido acetate
Synonyms: N-acetylacetamido acetate
Molecular Weight (g/mol): 159.14
Molecular Formula: C6H9NO4
InChi Key: NGULNJDDRUSDPR-UHFFFAOYSA-N
SMILES: CC(=O)ON(C(C)=O)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results