accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1773-44-0

IUPAC Name: (2E)-1-(2,4-dinitrophenyl)-2-[(2-methylphenyl)methylidene]hydrazine
Synonyms: (2E)-1-(2,4-dinitrophenyl)-2-[(2-methylphenyl)methylidene]hydrazine
Molecular Weight (g/mol): 300.27
Molecular Formula: C14H12N4O4
InChi Key: QUIFNJPMGQCNMH-OQLLNIDSSA-N
SMILES: CC1=CC=CC=C1\C=N\NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results