accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17745-45-8

IUPAC Name: propylboronic acid
Synonyms: propylboronic acid
Molecular Weight (g/mol): 87.91
Molecular Formula: C3H9BO2
InChi Key: JAQOMSTTXPGKTN-UHFFFAOYSA-N
SMILES: CCCB(O)O
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)