accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3OHO

CAS RN 17754-90-4

IUPAC Name: 4-(diethylamino)-2-hydroxybenzaldehyde
Synonyms: 4-(diethylamino)-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: XFVZSRRZZNLWBW-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=O)C(O)=C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (4)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)