accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17756-56-8

IUPAC Name: tetrahexylazanium hydroxide
Synonyms: tetrahexylazanium hydroxide
Molecular Weight (g/mol): 371.69
Molecular Formula: C24H53NO
InChi Key: JCJNUSDBRRKQPC-UHFFFAOYSA-M
SMILES: [OH-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results