accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHNH2

CAS RN 177583-27-6

IUPAC Name: tert-butyl N-[4-(aminomethyl)cyclohexyl]carbamate
Synonyms: tert-butyl N-[4-(aminomethyl)cyclohexyl]carbamate
Molecular Weight (g/mol): 228.34
Molecular Formula: C12H24N2O2
InChi Key: NVQFOBONHIXDOC-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CN)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)