accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONH2

CAS RN 1776-53-0

IUPAC Name: 4-aminocyclohexane-1-carboxylic acid
Synonyms: 4-aminocyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: DRNGLYHKYPNTEA-UHFFFAOYSA-N
SMILES: NC1CCC(CC1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)