accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17763-67-6

IUPAC Name: phenyl trifluoromethanesulfonate
Synonyms: phenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 226.17
Molecular Formula: C7H5F3O3S
InChi Key: GRJHONXDTNBDTC-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)