accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17763-91-6

IUPAC Name: 2-(trifluoromethanesulfonyloxy)phenyl trifluoromethanesulfonate
Synonyms: 2-(trifluoromethanesulfonyloxy)phenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 374.22
Molecular Formula: C8H4F6O6S2
InChi Key: XISAIQHXAICQIV-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC=CC=C1OS(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results