accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OBrHO

CAS RN 17764-93-1

IUPAC Name: 1-(5-bromo-2-hydroxyphenyl)propan-1-one
Synonyms: 1-(5-bromo-2-hydroxyphenyl)propan-1-one
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: NDXLYRKTHTZXRC-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC(Br)=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results