accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOHO

CAS RN 17768-28-4

IUPAC Name: 2-[3-(carboxymethyl)adamantan-1-yl]acetic acid
Synonyms: [3-(carboxymethyl)adamantan-1-yl]acetic acid
Molecular Weight (g/mol): 252.31
Molecular Formula: C14H20O4
InChi Key: UTENGZNBNPABQE-UHFFFAOYSA-N
SMILES: OC(=O)CC12CC3CC(C1)CC(CC(O)=O)(C3)C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results