accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17768-36-4

OH(R)(S)OO

CAS RN 17768-36-4

IUPAC Name: 2-[(5R,7S)-3-hydroxyadamantan-1-yl]acetate
Synonyms: 2-[(5R,7S)-3-hydroxyadamantan-1-yl]acetate
Molecular Weight (g/mol): 209.27
Molecular Formula: C12H17O3
InChi Key: JFMDWSCOQLUOCZ-LRSDLPTKSA-M
SMILES: OC12C[C@@H]3C[C@H](C1)CC(CC([O-])=O)(C3)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results