accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 177775-87-0

IUPAC Name: 9-benzoyl-3-bromo-9H-carbazole
Synonyms: 9-benzoyl-3-bromocarbazole
Molecular Weight (g/mol): 350.22
Molecular Formula: C19H12BrNO
InChi Key: CSLLZWBWBQIUIG-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)N(C(=O)C1=CC=CC=C1)C1=C2C=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results